Anti-JUN Mouse Monoclonal Antibody [clone: 7H5]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF800586EA 5700 DKK
ORIGCF800586
Anti-JUN Mouse Monoclonal Antibody [clone: 7H5]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: JUN
Clonality: Monoclonal
Clone: 7H5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human