Anti-CEP68 Mouse Monoclonal Antibody [clone: 6B4]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF507047EA 5870 DKK
ORIGCF507047
Anti-CEP68 Mouse Monoclonal Antibody [clone: 6B4]
Antistoffer
Recommended Dilutions: Western Blot: 1:400-4000
Type: Primary
Antigen: CEP68
Clonality: Monoclonal
Clone: 6B4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human