Anti-TMX1 Mouse Monoclonal Antibody [clone: 3H5]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF507042EA 5700 DKK
ORIGCF507042
Anti-TMX1 Mouse Monoclonal Antibody [clone: 3H5]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: TMX1
Clonality: Monoclonal
Clone: 3H5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR