Anti-MFAP3 Mouse Monoclonal Antibody [clone: 2D2]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF507040EA 5870 DKK
ORIGCF507040
Anti-MFAP3 Mouse Monoclonal Antibody [clone: 2D2]
Antistoffer
Recommended Dilutions: Western Blot: 1:1000-1:4000; Immunofluorescence: 1:100
Type: Primary
Antigen: MFAP3
Clonality: Monoclonal
Clone: 2D2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human