Anti-C17orf62 Mouse Monoclonal Antibody [clone: 3F10]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF507020EA 5700 DKK
ORIGCF507020
Anti-C17orf62 Mouse Monoclonal Antibody [clone: 3F10]
Antistoffer
Recommended Dilutions: Western Blot: 1:4000
Type: Primary
Antigen: C17orf62
Clonality: Monoclonal
Clone: 3F10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human