Anti-VIM Mouse Monoclonal Antibody [clone: 1F10]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF801359EA 5590 DKK
ORIGCF801359
Anti-VIM Mouse Monoclonal Antibody [clone: 1F10]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: 1F10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR