Anti-VIM Mouse Monoclonal Antibody [clone: 7B8]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF801358EA 5590 DKK
ORIGCF801358
Anti-VIM Mouse Monoclonal Antibody [clone: 7B8]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: 7B8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR