Anti-NUB1 Mouse Monoclonal Antibody [clone: 4H2]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF800188EA 5590 DKK
ORIGCF800188
Anti-NUB1 Mouse Monoclonal Antibody [clone: 4H2]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: NUB1
Clonality: Monoclonal
Clone: 4H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR