Anti-MSMB Mouse Monoclonal Antibody [clone: 2H1]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF803509EA 5590 DKK
ORIGCF803509
Anti-MSMB Mouse Monoclonal Antibody [clone: 2H1]
Antistoffer
Recommended Dilutions: Western Blot: 1:200-1:1000

Type: Primary
Antigen: MSMB
Clonality: Monoclonal
Clone: 2H1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR