Anti-FOXI1 Mouse Monoclonal Antibody [clone: 7G9]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF800141EA 5590 DKK
ORIGCF800141
Anti-FOXI1 Mouse Monoclonal Antibody [clone: 7G9]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: FOXI1
Clonality: Monoclonal
Clone: 7G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR