Anti-NR2C1 Mouse Monoclonal Antibody [clone: 1A9]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF803380EA 5750 DKK
ORIGCF803380
Anti-NR2C1 Mouse Monoclonal Antibody [clone: 1A9]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: NR2C1
Clonality: Monoclonal
Clone: 1A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR