Anti-GBA Mouse Monoclonal Antibody [clone: 4G4]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF803377EA 5590 DKK
ORIGCF803377
Anti-GBA Mouse Monoclonal Antibody [clone: 4G4]
Antistoffer
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150

Type: Primary
Antigen: GBA
Clonality: Monoclonal
Clone: 4G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR