Anti-NR2C1 Mouse Monoclonal Antibody [clone: 3C6]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF803359EA 5750 DKK
ORIGCF803359
Anti-NR2C1 Mouse Monoclonal Antibody [clone: 3C6]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: NR2C1
Clonality: Monoclonal
Clone: 3C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR