Anti-NKX2-1 Mouse Monoclonal Antibody [clone: 15E3]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF803320EA 5590 DKK
ORIGCF803320
Anti-NKX2-1 Mouse Monoclonal Antibody [clone: 15E3]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: NKX2-1
Clonality: Monoclonal
Clone: 15E3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR