Anti-INHA Mouse Monoclonal Antibody [clone: 12F11]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF801920EA 5590 DKK
ORIGCF801920
Anti-INHA Mouse Monoclonal Antibody [clone: 12F11]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: INHA
Clonality: Monoclonal
Clone: 12F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR