Anti-PRRX1 Mouse Monoclonal Antibody [clone: 4E11]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF803173EA 5590 DKK
ORIGCF803173
Anti-PRRX1 Mouse Monoclonal Antibody [clone: 4E11]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: PRRX1
Clonality: Monoclonal
Clone: 4E11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR