Anti-KI67 Mouse Monoclonal Antibody [clone: 18G5]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF803157EA 5590 DKK
ORIGCF803157
Anti-KI67 Mouse Monoclonal Antibody [clone: 18G5]
Antistoffer
Recommended Dilutions: Immunochemistry: 1:150

Type: Primary
Antigen: KI67
Clonality: Monoclonal
Clone: 18G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR