Anti-VIM Mouse Monoclonal Antibody [clone: 5B9]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF801121EA 5590 DKK
ORIGCF801121
Anti-VIM Mouse Monoclonal Antibody [clone: 5B9]
Antistoffer
Recommended Dilutions: Western Blot: 1:200-1:1000

Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: 5B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR