Anti-HNRNPM Mouse Monoclonal Antibody [clone: 3F3]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF803107EA 5590 DKK
ORIGCF803107
Anti-HNRNPM Mouse Monoclonal Antibody [clone: 3F3]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: HNRNPM
Clonality: Monoclonal
Clone: 3F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR