Anti-KLK8 Mouse Monoclonal Antibody [clone: 1G9]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF801720EA 5590 DKK
ORIGCF801720
Anti-KLK8 Mouse Monoclonal Antibody [clone: 1G9]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: KLK8
Clonality: Monoclonal
Clone: 1G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR