Anti-CDCA7L Mouse Monoclonal Antibody [clone: 3B5]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF803020EA 5590 DKK
ORIGCF803020
Anti-CDCA7L Mouse Monoclonal Antibody [clone: 3B5]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: CDCA7L
Clonality: Monoclonal
Clone: 3B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR