Anti-PCNA Mouse Monoclonal Antibody [clone: 3D6]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF800894EA 5750 DKK
ORIGCF800894
Anti-PCNA Mouse Monoclonal Antibody [clone: 3D6]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: PCNA
Clonality: Monoclonal
Clone: 3D6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR