Anti-C12orf26 Mouse Monoclonal Antibody [clone: 2B6]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF507335EA 5750 DKK
ORIGCF507335
Anti-C12orf26 Mouse Monoclonal Antibody [clone: 2B6]
Antistoffer
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150

Type: Primary
Antigen: C12orf26
Clonality: Monoclonal
Clone: 2B6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR