Anti-CD68 Mouse Monoclonal Antibody [clone: 12A3]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF802965EA 5590 DKK
ORIGCF802965
Anti-CD68 Mouse Monoclonal Antibody [clone: 12A3]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CD68
Clonality: Monoclonal
Clone: 12A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR