Anti-CDKN2A Mouse Monoclonal Antibody [clone: 1D3]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF802963EA 5590 DKK
ORIGCF802963
Anti-CDKN2A Mouse Monoclonal Antibody [clone: 1D3]
Antistoffer
Recommended Dilutions: Western Blot: 1:200-1:1000

Type: Primary
Antigen: CDKN2A
Clonality: Monoclonal
Clone: 1D3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR