Anti-NAB1 Mouse Monoclonal Antibody [clone: 5F2]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF802914EA 5590 DKK
ORIGCF802914
Anti-NAB1 Mouse Monoclonal Antibody [clone: 5F2]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: NAB1
Clonality: Monoclonal
Clone: 5F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR