Anti-CSH1 Mouse Monoclonal Antibody [clone: 2A2]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF506813EA 5590 DKK
ORIGCF506813
Anti-CSH1 Mouse Monoclonal Antibody [clone: 2A2]
Antistoffer
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100

Type: Primary
Antigen: CSH1
Clonality: Monoclonal
Clone: 2A2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR