Anti-MMP13 Mouse Monoclonal Antibody [clone: 4B7]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF506804EA 5590 DKK
ORIGCF506804
Anti-MMP13 Mouse Monoclonal Antibody [clone: 4B7]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: MMP13
Clonality: Monoclonal
Clone: 4B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR