Anti-CSH1 Mouse Monoclonal Antibody [clone: 2E8]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF506791EA 5590 DKK
ORIGCF506791
Anti-CSH1 Mouse Monoclonal Antibody [clone: 2E8]
Antistoffer
Recommended Dilutions: Western Blot: 1:4000
Type: Primary
Antigen: CSH1
Clonality: Monoclonal
Clone: 2E8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human