Anti-CD44 Mouse Monoclonal Antibody [clone: 5B5]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF506748EA 5590 DKK
ORIGCF506748
Anti-CD44 Mouse Monoclonal Antibody [clone: 5B5]
Antistoffer
Recommended Dilutions: Western Blot: 1:400-4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: 5B5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR