Anti-DPP3 Mouse Monoclonal Antibody [clone: 2D9]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF503242EA 5750 DKK
ORIGCF503242
Anti-DPP3 Mouse Monoclonal Antibody [clone: 2D9]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: DPP3
Clonality: Monoclonal
Clone: 2D9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR