Anti-PSMB7 Mouse Monoclonal Antibody [clone: 1D1]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF504429EA 5750 DKK
ORIGCF504429
Anti-PSMB7 Mouse Monoclonal Antibody [clone: 1D1]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: PSMB7
Clonality: Monoclonal
Clone: 1D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Monkey, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR