Anti-PRMT2 Mouse Monoclonal Antibody [clone: 1A3]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF503641EA 5750 DKK
ORIGCF503641
Anti-PRMT2 Mouse Monoclonal Antibody [clone: 1A3]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: PRMT2
Clonality: Monoclonal
Clone: 1A3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR