Anti-CRYAA Mouse Monoclonal Antibody [clone: 3B12]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF505577EA 5590 DKK
ORIGCF505577
Anti-CRYAA Mouse Monoclonal Antibody [clone: 3B12]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: CRYAA
Clonality: Monoclonal
Clone: 3B12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human