Anti-DIXDC1 Mouse Monoclonal Antibody [clone: 6B6]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF505550EA 5750 DKK
ORIGCF505550
Anti-DIXDC1 Mouse Monoclonal Antibody [clone: 6B6]
Antistoffer
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: DIXDC1
Clonality: Monoclonal
Clone: 6B6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR