Anti-TOMM34 Mouse Monoclonal Antibody [clone: 5E12]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF503182EA 5590 DKK
ORIGCF503182
Anti-TOMM34 Mouse Monoclonal Antibody [clone: 5E12]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TOMM34
Clonality: Monoclonal
Clone: 5E12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR