Anti-RUVBL2 Mouse Monoclonal Antibody [clone: 2B9]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF504384EA 5590 DKK
ORIGCF504384
Anti-RUVBL2 Mouse Monoclonal Antibody [clone: 2B9]
Antistoffer
Recommended Dilutions: Western Blot: 1:1000-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: RUVBL2
Clonality: Monoclonal
Clone: 2B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR