Anti-CDKN3 Mouse Monoclonal Antibody [clone: 2E11]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF503580EA 5590 DKK
ORIGCF503580
Anti-CDKN3 Mouse Monoclonal Antibody [clone: 2E11]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: CDKN3
Clonality: Monoclonal
Clone: 2E11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human