Anti-BTLA Mouse Monoclonal Antibody [clone: 1B4]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF505518EA 5590 DKK
ORIGCF505518
Anti-BTLA Mouse Monoclonal Antibody [clone: 1B4]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: BTLA
Clonality: Monoclonal
Clone: 1B4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human