Anti-C2orf62 Mouse Monoclonal Antibody [clone: 4E1]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF505506EA 5750 DKK
ORIGCF505506
Anti-C2orf62 Mouse Monoclonal Antibody [clone: 4E1]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: C2orf62
Clonality: Monoclonal
Clone: 4E1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR