Anti-TOMM34 Mouse Monoclonal Antibody [clone: 1B6]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF503119EA 5590 DKK
ORIGCF503119
Anti-TOMM34 Mouse Monoclonal Antibody [clone: 1B6]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: TOMM34
Clonality: Monoclonal
Clone: 1B6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR