Anti-LENG1 Mouse Monoclonal Antibody [clone: 1C6]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF503114EA 5750 DKK
ORIGCF503114
Anti-LENG1 Mouse Monoclonal Antibody [clone: 1C6]
Antistoffer
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: LENG1
Clonality: Monoclonal
Clone: 1C6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human