Anti-CYP2B6 Mouse Monoclonal Antibody [clone: 3D5]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF504328EA 5590 DKK
ORIGCF504328
Anti-CYP2B6 Mouse Monoclonal Antibody [clone: 3D5]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: CYP2B6
Clonality: Monoclonal
Clone: 3D5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Mouse, Rat