Anti-PDE4B Mouse Monoclonal Antibody [clone: 3A10]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF503532EA 5590 DKK
ORIGCF503532
Anti-PDE4B Mouse Monoclonal Antibody [clone: 3A10]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: PDE4B
Clonality: Monoclonal
Clone: 3A10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR