Anti-C2orf62 Mouse Monoclonal Antibody [clone: 2E6]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF505498EA 5750 DKK
ORIGCF505498
Anti-C2orf62 Mouse Monoclonal Antibody [clone: 2E6]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: C2orf62
Clonality: Monoclonal
Clone: 2E6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human