Anti-GADD45G Mouse Monoclonal Antibody [clone: B7]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF505477EA 5590 DKK
ORIGCF505477
Anti-GADD45G Mouse Monoclonal Antibody [clone: B7]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: GADD45G
Clonality: Monoclonal
Clone: B7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR