Anti-NDOR1 Mouse Monoclonal Antibody [clone: 1E3]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF505464EA 5590 DKK
ORIGCF505464
Anti-NDOR1 Mouse Monoclonal Antibody [clone: 1E3]
Antistoffer
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: NDOR1
Clonality: Monoclonal
Clone: 1E3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR