Anti-SH2B3 Mouse Monoclonal Antibody [clone: 1F6]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF503069EA 5590 DKK
ORIGCF503069
Anti-SH2B3 Mouse Monoclonal Antibody [clone: 1F6]
Antistoffer
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SH2B3
Clonality: Monoclonal
Clone: 1F6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR