Anti-UCHL1 Mouse Monoclonal Antibody [clone: 2B5]

Leverandør: OriGene
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ORIGCF504289EA 5590 DKK
ORIGCF504289
Anti-UCHL1 Mouse Monoclonal Antibody [clone: 2B5]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:50; Flow Cytometry: 1:100

Type: Primary
Antigen: UCHL1
Clonality: Monoclonal
Clone: 2B5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR