Anti-LGALS3BP Mouse Monoclonal Antibody [clone: 5E3]
Leverandør: OriGene
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ORIGCF503457EA 5590 DKK
ORIGCF503457
Anti-LGALS3BP Mouse Monoclonal Antibody [clone: 5E3]
Antistoffer
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100
Type: Primary
Antigen: LGALS3BP
Clonality: Monoclonal
Clone: 5E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human